ENA458425161 Enamine

4-cyanocyclohexane-1-carboxylic acid

  • MDL number MFCD20639432
  • Cas Number : 4848-16-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-105437

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-105437
Availability details for
Quantity
  • Availability
  • Add to Cart