ENA458425248 Enamine

4-{[3-(methoxymethyl)phenyl]amino}pyridine-2-carbonitrile

  • MDL number MFCD17408806
  • Cas Number : 1275856-40-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-223406

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223406
Availability details for
Quantity
  • Availability
  • Add to Cart