ENA458425270 Enamine

1-tert-butylpiperazine-2,5-dione

  • MDL number MFCD21115160
  • Cas Number : 1491454-34-2
  • Molecular Weight : 170.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-223473

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223473
Availability details for
Quantity
  • Availability
  • Add to Cart