ENA458425574 Enamine

3-methyl-1-[(thiophen-2-yl)methyl]-1H-pyrazol-5-amine hydrochloride

  • MDL number MFCD29033971
  • Cas Number : 1820618-78-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 85
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-224625

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224625
Availability details for
Quantity
  • Availability
  • Add to Cart