ENA458425596 Enamine

4-[(dimethylamino)methyl]-1,2-dimethyl-1H-1,3-benzodiazol-5-ol

  • MDL number MFCD29033999
  • Cas Number : 1702796-54-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-224912

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224912
Availability details for
Quantity
  • Availability
  • Add to Cart