ENA458425598 Enamine

4-{methyl[2-(methylamino)ethyl]amino}benzene-1-sulfonamide hydrochloride

  • MDL number MFCD29034001
  • Cas Number : 1820710-70-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-224952

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224952
Availability details for
Quantity
  • Availability
  • Add to Cart