ENA458425606 Enamine

3-bromo-5-fluoro-1-methyl-1H-pyrazole

  • MDL number MFCD29034014
  • Cas Number : 1820608-53-4
  • Molecular Weight : 178.99

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form liquid
Shipping Temperature Ice Pack
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-225043

Safety Information

signalword Danger
pcodes P210,P261,P264,P271,P280,P302+P352,P304+P340,P305+P351+P338,P310,P312,P332+P313,P362,P370+P378,P403+P233,P403+P235,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-225043
pictograms GHS05,GHS07
hstatements H227,H315,H318,H335
Availability details for
Quantity
  • Availability
  • Add to Cart