ENA458425633 Enamine

3,3,6-trimethyl-1-(methylsulfanyl)-3,4-dihydroisoquinoline

  • MDL number MFCD29034046
  • Cas Number : 1428732-23-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225300

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225300
Availability details for
Quantity
  • Availability
  • Add to Cart