ENA458425651 Enamine

ethyl 6-cyanopyrimidine-4-carboxylate

  • MDL number MFCD29034069
  • Cas Number : 1820617-97-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225554

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225554
Availability details for
Quantity
  • Availability
  • Add to Cart