ENA458425669 Enamine

methoxy[1-(2-methyl-1,3-thiazol-4-yl)ethylidene]amine

  • Cas Number : 1868663-01-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225696

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225696
Availability details for
Quantity
  • Availability
  • Add to Cart