ENA458425679 Enamine

3-hydroxy-8-methyl-8-(propan-2-yl)-8-azabicyclo[3.2.1]octan-8-ium iodide

  • MDL number MFCD29034101
  • Molecular Weight : 311.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (1R,5S,8r)-3-hydroxy-8-isopropyl-8-methyl-8-azabicyclo[3.2.1]octan-8-ium iodide
Inchi Code 1S/C11H22NO.HI/c1-8(2)12(3)9-4-5-10(12)7-11(13)6-9;/h8-11,13H,4-7H2,1-3H3;1H/q+1;/p-1
Inchi Key AMDWIUXLHSXQFJ-UHFFFAOYSA-M

Description

Purity 95
Salt Data [I-]
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-225842

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225842
Availability details for
Quantity
  • Availability
  • Add to Cart