ENA458425680 Enamine

4-nitro-2-(propan-2-yl)aniline hydrochloride

  • MDL number MFCD29034102
  • Cas Number : 1820685-08-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225854

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225854
Availability details for
Quantity
  • Availability
  • Add to Cart