ENA458425688 Enamine

3-bromo-4-chloro-5-fluoro-1-methyl-1H-pyrazole

  • MDL number MFCD29034112
  • Cas Number : 1820666-18-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226017

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226017
Availability details for
Quantity
  • Availability
  • Add to Cart