ENA458425696 Enamine

3-(aminomethyl)-N-(2,2,2-trifluoroethyl)benzene-1-sulfonamide hydrochloride

  • MDL number MFCD29034122
  • Cas Number : 1820716-71-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226084

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226084
Availability details for
Quantity
  • Availability
  • Add to Cart