ENA458425708 Enamine

2-[4-(1-aminoethyl)phenyl]-4-methyl-1lambda6,2-thiazolidine-1,1-dione hydrochloride

  • MDL number MFCD29034136
  • Cas Number : 1820718-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226214

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226214
Availability details for
Quantity
  • Availability
  • Add to Cart