ENA458425805 Enamine

4-(prop-2-yn-1-yl)morpholine-3,5-dione

  • MDL number MFCD14631778
  • Cas Number : 1248279-27-7
  • Molecular Weight : 153.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. +4
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-224831

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224831
Availability details for
Quantity
  • Availability
  • Add to Cart