ENA458425817 Enamine

3-iodo-6-(trifluoromethyl)pyridazine

  • MDL number MFCD18072596
  • Cas Number : 1206524-33-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-224850

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-224850
Availability details for
Quantity
  • Availability
  • Add to Cart