ENA458425910 Enamine

3,4-diamino-5-(trifluoromethyl)benzonitrile

  • MDL number MFCD24160440
  • Cas Number : 157554-49-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225104

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225104
Availability details for
Quantity
  • Availability
  • Add to Cart