ENA458425937 Enamine

1-(1-bromoethyl)-4-(trifluoromethyl)cyclohexane

  • MDL number MFCD21776284
  • Cas Number : 1536230-12-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225178

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225178
Availability details for
Quantity
  • Availability
  • Add to Cart