ENA458425938 Enamine

ethyl 1-(2-nitrophenyl)piperidine-2-carboxylate

  • MDL number MFCD11041913
  • Cas Number : 1255665-86-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-223925

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223925
Availability details for
Quantity
  • Availability
  • Add to Cart