ENA458425942 Enamine

4-amino-3-thiatricyclo[5.2.1.0,2,6]deca-2(6),4-diene-5-carbonitrile

  • MDL number MFCD24121282
  • Cas Number : 60948-80-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225184

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225184
Availability details for
Quantity
  • Availability
  • Add to Cart