ENA458426050 Enamine

7-chloro-3-methyl-1-benzothiophene-2-carboxylic acid

  • MDL number MFCD11104814
  • Cas Number : 752135-41-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225517

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225517
Availability details for
Quantity
  • Availability
  • Add to Cart