ENA458426078 Enamine

2-[3-(trifluoromethyl)phenyl]-3,4-dihydro-2H-1,4-benzoxazine

  • MDL number MFCD29034075
  • Cas Number : 1508099-32-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225603
Availability details for
Quantity
  • Availability
  • Add to Cart