ENA458426147 Enamine

1-[(methylamino)methyl]-1,2,3,4-tetrahydronaphthalen-1-ol

  • MDL number MFCD17218418
  • Cas Number : 105271-55-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-223548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223548
Availability details for
Quantity
  • Availability
  • Add to Cart