ENA458426151 Enamine

5-(2-methoxyphenyl)-1,3-oxazolidin-2-one

  • MDL number MFCD16303598
  • Cas Number : 1225523-75-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225838

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225838
Availability details for
Quantity
  • Availability
  • Add to Cart