ENA458426176 Enamine

5-bromo-N-(1-methyl-1H-pyrazol-4-yl)pyrimidin-2-amine

  • MDL number MFCD18902555
  • Cas Number : 1275374-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-225919

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225919
Availability details for
Quantity
  • Availability
  • Add to Cart