ENA458426178 Enamine

4-[3-amino-2-(trifluoromethyl)propyl]-1lambda6-thiomorpholine-1,1-dione

  • MDL number MFCD29033920
  • Cas Number : 1697949-22-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-223712

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223712
Availability details for
Quantity
  • Availability
  • Add to Cart