ENA458426184 Enamine

methyl 2-[(2-methylprop-2-en-1-yl)oxy]benzoate

  • MDL number MFCD29034106
  • Cas Number : 110124-06-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-225938

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-225938
Availability details for
Quantity
  • Availability
  • Add to Cart