ENA458426286 Enamine

1-(5-bromo-1H-indol-2-yl)ethan-1-amine

  • MDL number MFCD18655419
  • Cas Number : 1270476-46-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226209

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226209
Availability details for
Quantity
  • Availability
  • Add to Cart