ENA458426305 Enamine

tert-butyl 4-butanoylpiperazine-1-carboxylate

  • MDL number MFCD16892450
  • Cas Number : 913952-85-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226295

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226295
Availability details for
Quantity
  • Availability
  • Add to Cart