ENA458426353 Enamine

7-bromo-6-chloro-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD29034156
  • Cas Number : 1700096-22-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226386

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226386
Availability details for
Quantity
  • Availability
  • Add to Cart