ENA458426379 Enamine

N-(2-bromo-5-chlorophenyl)-3-phenylprop-2-enamide

  • Cas Number : 1563516-59-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226432

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226432
Availability details for
Quantity
  • Availability
  • Add to Cart