ENA458426458 Enamine

2-(cyclohex-1-en-1-yl)pyridine

  • MDL number MFCD06637511
  • Cas Number : 14159-55-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-226591

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226591
Availability details for
Quantity
  • Availability
  • Add to Cart