ENA458426605 Enamine

1-[2-(2-fluorophenyl)-1,3-thiazol-4-yl]ethan-1-amine hydrochloride

  • MDL number MFCD29034237
  • Cas Number : 1820706-03-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Storage temp. Normal
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-227215

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227215
Availability details for
Quantity
  • Availability
  • Add to Cart