ENA458426632 Enamine

2-chloro-5-(2-methoxyphenyl)-1,3,4-thiadiazole

  • MDL number MFCD11845082
  • Cas Number : 40288-19-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226914

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226914
Availability details for
Quantity
  • Availability
  • Add to Cart