ENA458426646 Enamine

1-ethylazetidin-3-amine

  • MDL number MFCD18817602
  • Cas Number : 55438-58-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Ice Pack
Product Link https://www.enaminestore.com/catalog/EN300-226954

Safety Information

signalword Danger
pcodes P260,P264,P270,P280,P301+P312,P301+P330+P331,P302+P352,P303+P361+P353,P304+P340,P305+P351+P338,P310,P312,P330,P363,P405,P501
hcodes C,Xn
MSDS https://www.enaminestore.com/catalog/EN300-226954
pictograms GHS05,GHS07
hstatements H302,H312,H314
Availability details for
Quantity
  • Availability
  • Add to Cart