ENA458426692 Enamine

N-(2-methoxy-5-methylphenyl)-2-methyl-6-(3-methylphenyl)pyrimidin-4-amine

  • MDL number MFCD14831899
  • Cas Number : 1019132-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226820

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226820
Availability details for
Quantity
  • Availability
  • Add to Cart