ENA458426693 Enamine

2-(4-bromo-3-fluorophenyl)oxirane

  • MDL number MFCD24108103
  • Cas Number : 1400997-35-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226823

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226823
Availability details for
Quantity
  • Availability
  • Add to Cart