ENA458426779 Enamine

2-(benzyloxy)-4-chloroaniline

  • MDL number MFCD16732087
  • Cas Number : 860677-88-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Normal
Shipping Temperature Normal
Product Link https://www.enaminestore.com/catalog/EN300-226878

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-226878
Availability details for
Quantity
  • Availability
  • Add to Cart