ENA458426959 Enamine

N-[(1,3-dimethyl-1H-pyrazol-4-yl)methylidene]hydroxylamine

  • Cas Number : 380583-07-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-227748

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227748
Availability details for
Quantity
  • Availability
  • Add to Cart