ENA458426966 Enamine

3-({[4-(difluoromethyl)-6-methylpyrimidin-2-yl]sulfanyl}methyl)-4-methoxybenzaldehyde

  • MDL number MFCD02253745
  • Cas Number : 438218-64-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-227755

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227755
Availability details for
Quantity
  • Availability
  • Add to Cart