ENA458427024 Enamine

methyl 5-[(4-ethylphenoxy)methyl]furan-2-carboxylate

  • MDL number MFCD02253905
  • Cas Number : 402772-24-1
  • Molecular Weight : 260.29

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-227813

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227813
Availability details for
Quantity
  • Availability
  • Add to Cart