ENA458427106 Enamine

4-bromo-1-[(3-methylphenyl)methyl]-1H-pyrazol-3-amine

  • MDL number MFCD02254034
  • Cas Number : 1004451-94-8
  • Molecular Weight : 266.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-227895

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227895
Availability details for
Quantity
  • Availability
  • Add to Cart