ENA458427204 Enamine

3-bromo-4-butoxy-5-ethoxybenzaldehyde

  • MDL number MFCD02256593
  • Cas Number : 881601-64-5
  • Molecular Weight : 301.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-227993

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-227993
Availability details for
Quantity
  • Availability
  • Add to Cart