ENA458427294 Enamine

4-butoxy-3-iodo-5-methoxybenzaldehyde

  • MDL number MFCD02257153
  • Cas Number : 834914-06-6
  • Molecular Weight : 334.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228083
Availability details for
Quantity
  • Availability
  • Add to Cart