ENA458427345 Enamine

5-bromo-3-iodo-2-(prop-2-en-1-yloxy)benzaldehyde

  • MDL number MFCD02257510
  • Cas Number : 832674-00-7
  • Molecular Weight : 366.98

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228134

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228134
Availability details for
Quantity
  • Availability
  • Add to Cart