ENA458427362 Enamine

2-(4-bromo-2-formylphenoxy)-N-(2-phenylethyl)acetamide

  • MDL number MFCD02257617
  • Cas Number : 692279-90-6
  • Molecular Weight : 362.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228151

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228151
Availability details for
Quantity
  • Availability
  • Add to Cart