ENA458427364 Enamine

2-(4-bromo-2-formylphenoxy)-N-(2-methylphenyl)acetamide

  • MDL number MFCD02257629
  • Cas Number : 553639-44-4
  • Molecular Weight : 348.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228153

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228153
Availability details for
Quantity
  • Availability
  • Add to Cart