ENA458427442 Enamine

3-[(4-chloro-3-nitro-1H-pyrazol-1-yl)methyl]-4-methoxybenzaldehyde

  • MDL number MFCD01114957
  • Cas Number : 514801-13-9
  • Molecular Weight : 295.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228231

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228231
Availability details for
Quantity
  • Availability
  • Add to Cart