ENA458427454 Enamine

1-(1,3-dimethyl-1H-pyrazol-4-yl)propan-2-amine

  • MDL number MFCD03419356
  • Cas Number : 1006357-26-1
  • Molecular Weight : 153.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-228243

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-228243
Availability details for
Quantity
  • Availability
  • Add to Cart